RIE156330493 Rieke Metals, Inc.

ethyl 6-(2-ethoxyphenyl)-6-oxohexanoate

  • MDL number MFCD02261415
  • Cas Number : 898757-42-1
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7654D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7654D
Availability details for
Quantity
  • Availability
  • Add to Cart