RIE156330494 Rieke Metals, Inc.

ethyl 7-(2-ethoxyphenyl)-7-oxoheptanoate

  • MDL number MFCD02261416
  • Cas Number : 898757-45-4
  • Molecular Weight : 292.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7654E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7654E
Availability details for
Quantity
  • Availability
  • Add to Cart