RIE156330497 Rieke Metals, Inc.

ethyl 7-(4-ethoxyphenyl)-7-oxoheptanoate

  • MDL number MFCD02261420
  • Cas Number : 898757-54-5
  • Molecular Weight : 292.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7655E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7655E
Availability details for
Quantity
  • Availability
  • Add to Cart