RIE156330500 Rieke Metals, Inc.

ethyl 5-oxo-5-(4-n-propoxyphenyl)valerate

  • MDL number MFCD02261422
  • Cas Number : 898757-60-3
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7656C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7656C
Availability details for
Quantity
  • Availability
  • Add to Cart