RIE156330501 Rieke Metals, Inc.

Ethyl 6-oxo-6-(4-n-propoxyphenyl)hexanoate

  • MDL number MFCD02261423
  • Cas Number : 898757-63-6
  • Molecular Weight : 292.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7656D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7656D
Availability details for
Quantity
  • Availability
  • Add to Cart