RIE156330505 Rieke Metals, Inc.

Ethyl 5-oxo-5-(4-isopropoxyphenyl)valerate

  • MDL number MFCD02261427
  • Cas Number : 898757-71-6
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7657C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7657C
Availability details for
Quantity
  • Availability
  • Add to Cart