RIE156330507 Rieke Metals, Inc.

Ethyl 7-oxo-7-(4-isopropoxyphenyl)heptanoate

  • MDL number MFCD02261429
  • Cas Number : 898757-75-0
  • Molecular Weight : 306.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7657E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7657E
Availability details for
Quantity
  • Availability
  • Add to Cart