RIE156330518 Rieke Metals, Inc.

ethyl 8-(4-hexyloxyphenyl)-8-oxooctanoate

  • MDL number MFCD02261443
  • Cas Number : 898757-95-4
  • Molecular Weight : 362.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7660F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7660F
Availability details for
Quantity
  • Availability
  • Add to Cart