RIE156330519 Rieke Metals, Inc.

Ethyl 4-(4-heptyloxyphenyl)-4-oxobutyrate

  • MDL number MFCD02261464
  • Cas Number : 143134-90-1
  • Molecular Weight : 320.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7661B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7661B
Availability details for
Quantity
  • Availability
  • Add to Cart