RIE156330521 Rieke Metals, Inc.

ethyl 6-(4-heptyloxyphenyl)-6-oxohexanoate

  • MDL number MFCD02261468
  • Cas Number : 898757-99-8
  • Molecular Weight : 348.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7661D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7661D
Availability details for
Quantity
  • Availability
  • Add to Cart