RIE156330523 Rieke Metals, Inc.

ethyl 8-(4-heptyloxyphenyl)-8-oxooctanoate

  • MDL number MFCD02261472
  • Cas Number : 898758-03-7
  • Molecular Weight : 376.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7661F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7661F
Availability details for
Quantity
  • Availability
  • Add to Cart