RIE156330534 Rieke Metals, Inc.

ethyl 4-(2,5-dimethoxyphenyl)-4-oxobutyrate

  • MDL number MFCD02261473
  • Cas Number : 898758-26-4
  • Molecular Weight : 266.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7667B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7667B
Availability details for
Quantity
  • Availability
  • Add to Cart