RIE156330536 Rieke Metals, Inc.

ethyl 6-(2,5-dimethoxyphenyl)-6-oxohexanoate

  • MDL number MFCD02261475
  • Cas Number : 898758-32-2
  • Molecular Weight : 294.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7667D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7667D
Availability details for
Quantity
  • Availability
  • Add to Cart