RIE156330537 Rieke Metals, Inc.

ethyl 7-(2,5-dimethoxyphenyl)-7-oxoheptanoate

  • MDL number MFCD02261476
  • Cas Number : 898758-35-5
  • Molecular Weight : 308.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7667E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7667E
Availability details for
Quantity
  • Availability
  • Add to Cart