RIE156330539 Rieke Metals, Inc.

ethyl 4-(2,6-dimethoxyphenyl)-4-oxobutyrate

  • MDL number MFCD02261478
  • Cas Number : 898758-41-3
  • Molecular Weight : 266.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7668B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7668B
Availability details for
Quantity
  • Availability
  • Add to Cart