RIE156330540 Rieke Metals, Inc.

ethyl 5-(2,6-dimethoxyphenyl)-5-oxovalerate

  • MDL number MFCD02261479
  • Cas Number : 898758-44-6
  • Molecular Weight : 280.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7668C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7668C
Availability details for
Quantity
  • Availability
  • Add to Cart