RIE156330542 Rieke Metals, Inc.

ethyl 7-(2,6-dimethoxyphenyl)-7-oxoheptanoate

  • MDL number MFCD02261481
  • Cas Number : 898758-50-4
  • Molecular Weight : 308.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7668E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7668E
Availability details for
Quantity
  • Availability
  • Add to Cart