RIE156330545 Rieke Metals, Inc.

ethyl 8-(3,4-dimethoxyphenyl)-8-oxooctanoate

  • MDL number MFCD02261486
  • Cas Number : 57641-19-7
  • Molecular Weight : 322.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7669F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7669F
Availability details for
Quantity
  • Availability
  • Add to Cart