RIE156330547 Rieke Metals, Inc.

ethyl 5-(3,5-dimethoxyphenyl)-5-oxovalerate

  • MDL number MFCD02261488
  • Cas Number : 898758-62-8
  • Molecular Weight : 280.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7670C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7670C
Availability details for
Quantity
  • Availability
  • Add to Cart