RIE156330552 Rieke Metals, Inc.

Ethyl 5-(2-acetoxyphenyl)-5-oxovalerate

  • MDL number MFCD07700170
  • Cas Number : 898758-73-1
  • Molecular Weight : 278.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7671C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7671C
Availability details for
Quantity
  • Availability
  • Add to Cart