RIE156330553 Rieke Metals, Inc.

Ethyl 6-(2-acetoxyphenyl)-6-oxohexanoate

  • MDL number MFCD07700171
  • Cas Number : 898758-75-3
  • Molecular Weight : 292.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7671D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7671D
Availability details for
Quantity
  • Availability
  • Add to Cart