RIE156330560 Rieke Metals, Inc.

Ethyl 4-(4-methoxy-3-nitrophenyl)-4-oxobutyrate

  • MDL number MFCD02261557
  • Cas Number : 898758-89-9
  • Molecular Weight : 281.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7685B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7685B
Availability details for
Quantity
  • Availability
  • Add to Cart