RIE156330562 Rieke Metals, Inc.

Ethyl 6-(4-methoxy-3-nitrophenyl)-6-oxohexanoate

  • MDL number MFCD02261559
  • Cas Number : 898758-93-5
  • Molecular Weight : 309.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7685D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7685D
Availability details for
Quantity
  • Availability
  • Add to Cart