RIE156330563 Rieke Metals, Inc.

Ethyl 7-(4-methoxy-3-nitrophenyl)-7-oxoheptanoate

  • MDL number MFCD02261560
  • Cas Number : 898758-95-7
  • Molecular Weight : 323.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7685E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7685E
Availability details for
Quantity
  • Availability
  • Add to Cart