RIE156330564 Rieke Metals, Inc.

Ethyl 8-(4-methoxy-3-nitrophenyl)-8-oxooctanoate

  • MDL number MFCD02261561
  • Cas Number : 898758-97-9
  • Molecular Weight : 337.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7685F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7685F
Availability details for
Quantity
  • Availability
  • Add to Cart