RIE156330565 Rieke Metals, Inc.

Ethyl 4-(2-methyl-5-fluorophenyl)-4-oxobutyrate

  • MDL number MFCD02261602
  • Cas Number : 898758-99-1
  • Molecular Weight : 238.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7697B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7697B
Availability details for
Quantity
  • Availability
  • Add to Cart