RIE156330566 Rieke Metals, Inc.

Ethyl 4-[3-(N,N-Dimethylamino)phenyl]-4-oxobutanoate

  • MDL number MFCD09801899
  • Molecular Weight : 249.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7699B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7699B
Availability details for
Quantity
  • Availability
  • Add to Cart