RIE156330569 Rieke Metals, Inc.

Ethyl 7-[3-(N,N-Dimethylamino)phenyl]-7-oxoheptanoate

  • MDL number MFCD09801902
  • Molecular Weight : 291.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7699E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7699E
Availability details for
Quantity
  • Availability
  • Add to Cart