RIE156330571 Rieke Metals, Inc.

Ethyl 5-[4-(N,N-Dimethylamino)phenyl]-5-oxovalerate

  • MDL number MFCD09801905
  • Molecular Weight : 263.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7700C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7700C
Availability details for
Quantity
  • Availability
  • Add to Cart