RIE156330574 Rieke Metals, Inc.

Ethyl 8-[4-(N,N-Dimethylamino)phenyl]-8-oxooctanoate

  • MDL number MFCD09801908
  • Molecular Weight : 305.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7700F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7700F
Availability details for
Quantity
  • Availability
  • Add to Cart