RIE156330577 Rieke Metals, Inc.

Ethyl 6-[4-(N,N-Diethylamino)phenyl]-6-oxohexanoate

  • MDL number MFCD09801911
  • Molecular Weight : 305.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7701D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7701D
Availability details for
Quantity
  • Availability
  • Add to Cart