RIE156330579 Rieke Metals, Inc.

Ethyl 8-[4-(N,N-Diethylamino)phenyl]-8-oxooctanoate

  • MDL number MFCD09801913
  • Molecular Weight : 333.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7701F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7701F
Availability details for
Quantity
  • Availability
  • Add to Cart