RIE156330580 Rieke Metals, Inc.

Ethyl 4-(2-Chloro-4-fluorophenyl)-4-oxobutanoate

  • MDL number MFCD09801914
  • Molecular Weight : 258.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7702B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7702B
Availability details for
Quantity
  • Availability
  • Add to Cart