RIE156330583 Rieke Metals, Inc.

Ethyl 7-(2-Chloro-4-fluorophenyl)-7-oxoheptanoate

  • MDL number MFCD09801917
  • Molecular Weight : 300.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7702E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7702E
Availability details for
Quantity
  • Availability
  • Add to Cart