RIE156330586 Rieke Metals, Inc.

Ethyl 5-(4-Chloro-2-fluorophenyl)-5-oxovalerate

  • MDL number MFCD09801920
  • Molecular Weight : 272.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7703C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7703C
Availability details for
Quantity
  • Availability
  • Add to Cart