RIE156330588 Rieke Metals, Inc.

Ethyl 7-(4-Chloro-2-fluorophenyl)-7-oxoheptanoate

  • MDL number MFCD09801922
  • Molecular Weight : 300.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7703E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7703E
Availability details for
Quantity
  • Availability
  • Add to Cart