RIE156330589 Rieke Metals, Inc.

Ethyl 8-(4-Chloro-2-fluorophenyl)-8-oxooctanoate

  • MDL number MFCD09801923
  • Molecular Weight : 314.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7703F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7703F
Availability details for
Quantity
  • Availability
  • Add to Cart