RIE156330597 Rieke Metals, Inc.

Ethyl 6-(5-Chloro-2-methoxyphenyl)-6-oxohexanoate

  • MDL number MFCD09801931
  • Molecular Weight : 298.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7705D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7705D
Availability details for
Quantity
  • Availability
  • Add to Cart