RIE156330602 Rieke Metals, Inc.

Ethyl 6-(2-Chloro-4-(methylthio)phenyl)-6-oxohexanoate

  • MDL number MFCD09801936
  • Molecular Weight : 314.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7706D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7706D
Availability details for
Quantity
  • Availability
  • Add to Cart