RIE156330603 Rieke Metals, Inc.

Ethyl 7-(2-Chloro-4-(methylthio)phenyl)-7-oxoheptanoate

  • MDL number MFCD09801937
  • Molecular Weight : 328.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7706E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7706E
Availability details for
Quantity
  • Availability
  • Add to Cart