RIE156330609 Rieke Metals, Inc.

Ethyl 8-(2-Chloro-6-fluorophenyl)-8-oxooctanoate

  • MDL number MFCD09801943
  • Molecular Weight : 314.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7707F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7707F
Availability details for
Quantity
  • Availability
  • Add to Cart