RIE156330616 Rieke Metals, Inc.

Ethyl 5-(2,3,4,5-tetrafluorophenyl)-5-oxovalerate

  • MDL number MFCD02261578
  • Molecular Weight : 292.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7709C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7709C
Availability details for
Quantity
  • Availability
  • Add to Cart