RIE156330617 Rieke Metals, Inc.

Ethyl 6-(2,3,4,5-tetrafluorophenyl)-6-oxohexanoate

  • MDL number MFCD02261579
  • Molecular Weight : 306.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7709D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7709D
Availability details for
Quantity
  • Availability
  • Add to Cart