RIE156330619 Rieke Metals, Inc.

Ethyl 8-(2,3,4,5-tetrafluorophenyl)-8-oxooctanoate

  • MDL number MFCD02261581
  • Molecular Weight : 334.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7709F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7709F
Availability details for
Quantity
  • Availability
  • Add to Cart