RIE156330620 Rieke Metals, Inc.

Ethyl 4-(3,4,5-trichlorophenyl)-4-oxobutanoate

  • MDL number MFCD09801949
  • Molecular Weight : 309.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7710B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7710B
Availability details for
Quantity
  • Availability
  • Add to Cart