RIE156330623 Rieke Metals, Inc.

Ethyl 7-(3,4,5-trichlorophenyl)-7-oxoheptanoate

  • MDL number MFCD09801952
  • Molecular Weight : 351.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7710E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7710E
Availability details for
Quantity
  • Availability
  • Add to Cart