RIE156330624 Rieke Metals, Inc.

Ethyl 8-(3,4,5-trichlorophenyl)-8-oxooctanoate

  • MDL number MFCD09801953
  • Molecular Weight : 365.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7710F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7710F
Availability details for
Quantity
  • Availability
  • Add to Cart