RIE156330630 Rieke Metals, Inc.

Ethyl 4-(2,4,5-trimethoxyphenyl)-4-oxobutanoate

  • MDL number MFCD09801959
  • Molecular Weight : 296.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7712B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7712B
Availability details for
Quantity
  • Availability
  • Add to Cart