RIE156330632 Rieke Metals, Inc.

Ethyl 6-(2,4,5-trimethoxyphenyl)-6-oxohexanoate

  • MDL number MFCD09801961
  • Molecular Weight : 324.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7712D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7712D
Availability details for
Quantity
  • Availability
  • Add to Cart