RIE156330633 Rieke Metals, Inc.

Ethyl 7-(2,4,5-trimethoxyphenyl)-7-oxoheptanoate

  • MDL number MFCD09801962
  • Molecular Weight : 338.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7712E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7712E
Availability details for
Quantity
  • Availability
  • Add to Cart